Learn More
Rockland Immunochemicals Molecular Biology Grade UltraPure Water (1 x 1L) - MB-010-1000
SDP
Catalog No. 501058975
Encompass_Preferred
Molecular Biology Grade UltraPure Water is free of all RNase enzymes. DEPC-treated (RNase-free) water is used in handling of RNA in the laboratory to reduce risks of RNA degradation by RNases. It is ideal for all molecular biology applications and is suitable for work involving DNA and RNA.
Catalog No. 50-105-8975 Supplier Rockland Immunochemicals Supplier No. MB0101000
Molecular Biology Grade UltraPure Water is free of all RNase enzymes. DEPC-treated (RNase-free) water is used in handling of RNA in the laboratory to reduce risks of RNA degradation by RNases. It is ideal for all molecular biology applications and is suitable for work involving DNA and RNA.