Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Click-iT™ Plus OPP Alexa Fluor™ 594 Protein Synthesis Assay Kit
GREENER_CHOICE

Catalog No. C10457
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 Kit
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C10457 1 Kit
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. C10457 Supplier Invitrogen™ Supplier No. C10457
  • $996.00
    $926.65 / Each of 1

    Save $69.35

    7% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
View all promotions
Add to Cart
Edge
Add to Cart

missing translation for 'validMainframeMsg'

Includes

Click-iT OPP Reagent, Alexa Fluor dye picolyl azide, Click-iT OPP Reaction Buffer, Copper Protectant, Click-iT Reaction Buffer Additive, Click-iT Reaction Rise Buffer and NuclearMask™ Blue Stain

Provides non-radioactive method for detection of protein synthesis using fluorescence microscopy or high-content imaging

The Click-iT™ Plus OPP Alexa Fluor™ 594 Protein Synthesis Assay Kit provides a fast, sensitive, and non-radioactive method for the detection of protein synthesis using fluorescence microscopy or high-content imaging. In this assay O-propargyl-puromycin (OPP) is efficiently incorporated into newly translated proteins in complete methionine-containing media and fluorescently labeled with a bright, photostable Alexa Fluor™ dye in a fast, highly-specific, and mild click reaction.

Features of the kit include:

  • No media change required – works in complete, methionine-containing media, no need to remove cell media
  • Multiplex-enabled – Click-iT™ Plus technology retains signal from GFP and binding of fluorescent-conjugated phalloidins
  • Non-radioactive – an alternative to the traditional 35S-methionine methods
  • Works in vivo—published results demonstrate use in vivo for determination of protein translation

The kit contains O-propargyl-puromycin (OPP), which is an alkyne analog of puromycin (also available separately), as well as Alexa Fluor™ 594 picolyl azide and all necessary reagents to perform the Click reaction. The OPP is fed to cultured cells and incorporated into proteins during active protein synthesis. Addition of the Alexa Fluor™ 594 picolyl azide and the Click reaction reagents leads to a chemoselective ligation, or 'click' reaction, between the picolyl azide dye and the OPP alkyne, allowing the modified proteins to be detected by imaged-based analysis.

The click reaction uses bioorthogonal (biologically unique) moieties to fluorescently label proliferating cells, helping to produce low backgrounds and high detection sensitivities. Because of the mild reaction conditions, Click-iT™ Plus assays detect protein translation events while enabling preservation of cell morphology, the binding of fluorescently-labeled phalloidin, and the fluorescent signal from GFP.

Unlike 35S-methionine, used in traditional methods, OPP is not an amino acid analog, so it can be added directly to cells in complete media or used to determine protein synthesis in vivo.

The kit contains all of the components needed to label and detect the incorporated OPP in newly translated proteins in samples of adherent cells. The kit includes sufficient reagents for the labeling of 25 18 mm × 18 mm coverslips using 1 mL of reaction buffer per test.

TRUSTED_SUSTAINABILITY

Specifications

For Use With (Application) Detection of protein synthesis using fluorescence microscopy or high-content imaging
Product Type Click-iT™ Plus OPP Alexa Fluor™ 594 Protein Synthesis Assay Kit
Quantity 1 Kit
Content And Storage This kit contains: Click-iT™ OPP Reagent, Alexa Fluor™ dye picolyl azide, Click-iT™ OPP Reaction Buffer, Copper Protectant, Click-iT™ Reaction Buffer Additive, Click-iT™ Reaction Rise Buffer, and NuclearMask™ Blue Stain. Store at 2° to 8°C. Desiccate and protect from light.
Reagent Type Nascent Protein Labeling Reagent
Product Line Click-iT
Label or Dye Alexa Fluor 594
I will be performing a cell proliferation assay using Click-iT EdU kit. At what point can I stop overnight, or do I have to perform all the steps continuously?

One may store the sample after fixation overnight in PBS at 4oC. For longer storage (<1 week) , store in buffer with 1-2% formaldehyde or in formalin to limit microbial growth. If you use sodium azide as a microbial inhibitor, it must be completely removed prior to the Click-iT reaction.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.