Learn More
TARGETMOL CHEMICALS INC MOLIBRESIB 10MG
SDP
Catalog No. 502577278
Encompass_Preferred
Also available in 1 mg 2 mg 5 mg 25 mg 50 mg 100 mg 200 mg 500 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. Molibresib (GSK525762) is an inhibitor of BET proteins (IC50 about 35 nM). purity: 99%
Catalog No. 50-257-7278 Supplier TARGETMOL CHEMICALS INC Supplier No. T190610MG
Also available in 1 mg 2 mg 5 mg 25 mg 50 mg 100 mg 200 mg 500 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. Molibresib (GSK525762) is an inhibitor of BET proteins (IC50 about 35 nM). purity: 99%