Learn More
Medchemexpress LLC Molibresib | 1260907-17-2 | 200 MG
SDP
Catalog No. 5000339134
Encompass_Preferred
Molibresib (I-BET762; GSK525762) is a BET bromodomain inhibitor with an IC₅₀ of 32.5-42.5 nM. It binds to the tandem bromodomains of BET with high affinity and displaces a tetra-acetylated H4 peptide. It has high affinity for the BD1/BD2 domain of BRD2/3/4 proteins and inhibits OPM-2 cell proliferation.
  • BET bromodomain inhibitor
  • High affinity for BD1/BD2 domain of BRD2/3/4 proteins
  • Inhibits OPM-2 cell proliferation
Catalog No. 50-003-39134 Supplier Medchemexpress LLC Supplier No. HY13032200MG
Molibresib (I-BET762; GSK525762) is a BET bromodomain inhibitor with an IC₅₀ of 32.5-42.5 nM. It binds to the tandem bromodomains of BET with high affinity and displaces a tetra-acetylated H4 peptide. It has high affinity for the BD1/BD2 domain of BRD2/3/4 proteins and inhibits OPM-2 cell proliferation.
  • BET bromodomain inhibitor
  • High affinity for BD1/BD2 domain of BRD2/3/4 proteins
  • Inhibits OPM-2 cell proliferation