Learn More
Medchemexpress LLC Molibresib besylate | 1895049-20-3 | 97.0% | 100 MG
SDP
Catalog No. 5000341039
Encompass_Preferred
Molibresib besylate is an orally active pan-BET inhibitor. It targets BRD2, BRD3, BRD4, and BRDT, disrupting their interaction with chromatin by occupying acetylated lysine binding sites. This inhibits MYC expression and target gene transcription. It exhibits broad antiproliferative activity, useful for research in acute myeloid leukemia, multiple myeloma, triple-negative breast cancer, small-cell lung cancer, and other advanced refractory solid tumors.
  • Disrupts BET protein-chromatin interaction
  • Inhibits MYC expression and target gene transcription
  • Exhibits broad antiproliferative activity
  • Induces growth arrest and downregulates mitosis-related genes
  • Upregulates p-ERK1/2 levels
  • Shows synergistic effects with MEK inhibitors
  • Modulates immune microenvironment and prolongs survival in models
Catalog No. 50-003-41039 Supplier Medchemexpress LLC Supplier No. HY13032B100MG
Molibresib besylate is an orally active pan-BET inhibitor. It targets BRD2, BRD3, BRD4, and BRDT, disrupting their interaction with chromatin by occupying acetylated lysine binding sites. This inhibits MYC expression and target gene transcription. It exhibits broad antiproliferative activity, useful for research in acute myeloid leukemia, multiple myeloma, triple-negative breast cancer, small-cell lung cancer, and other advanced refractory solid tumors.
  • Disrupts BET protein-chromatin interaction
  • Inhibits MYC expression and target gene transcription
  • Exhibits broad antiproliferative activity
  • Induces growth arrest and downregulates mitosis-related genes
  • Upregulates p-ERK1/2 levels
  • Shows synergistic effects with MEK inhibitors
  • Modulates immune microenvironment and prolongs survival in models