Learn More
Medchemexpress LLC HY-13032B 5mg Medchemexpress, Molibresib (besylate) CAS:1895049-20-3 Purity:>98%
SDP
Catalog No. 501932648
Encompass_Preferred
Medchemexpress, HY-13032B 5mg Molibresib (besylate) CAS:1895049-20-3 Molibresib besylate (GSK 525762C; I-BET 762 besylate) is a BET bromodomain inhibitor with IC 50 of 32.5-42.5 nM. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-193-2648 Supplier Medchemexpress LLC Supplier No. HY13032B5MG
Medchemexpress, HY-13032B 5mg Molibresib (besylate) CAS:1895049-20-3 Molibresib besylate (GSK 525762C; I-BET 762 besylate) is a BET bromodomain inhibitor with IC 50 of 32.5-42.5 nM. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.