Learn More
Selleck Chemical LLC Molidustat (BAY 85-3934) 5mg 1154028-82-6
SDP
Catalog No. 501365707
Encompass_Preferred
Molidustat (BAY 85-3934) is a potent hypoxia-inducible factor prolyl hydroxylase (HIF-PH) inhibitor with IC50 of 480 nM, 280 nM, and 450 nM for PHD1, PHD2, and PHD, respectively. Phase 2. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-136-5707 Supplier Selleck Chemical LLC Supplier No. S81385MG
Molidustat (BAY 85-3934) is a potent hypoxia-inducible factor prolyl hydroxylase (HIF-PH) inhibitor with IC50 of 480 nM, 280 nM, and 450 nM for PHD1, PHD2, and PHD, respectively. Phase 2. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.