Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Molindone-d8 | 1189805-13-7 | 99.2% | 284.42 g·mol⁻¹ | C16H16D8N2O2 | 1 MG
SDP

Catalog No. 5000241470
Encompass_Preferred

Molindone-d8 is the deuterium-labeled isotopologue of molindone supplied as a research-grade stable isotope standard for analytical and preclinical studies. It is intended for use as an internal standard and tracer in mass spectrometry, metabolic, and pharmacokinetic research.

  • Deuterium-labeled analog for isotope-dilution assays.
  • High purity suitable for analytical use (99.2%).
  • Molecular formula C16H16D8N2O2 and molecular weight 284.42 g·mol⁻¹.
  • Offered in small quantities suitable for trace-level studies (1 mg, 5 mg, 10 mg).
  • Recommended storage: powder -20 °C for long term; in solvent -80 °C for short term.
  • Common applications: LC-MS/MS internal standard, metabolic tracing, and pharmacokinetic studies.

Catalog No. 50-002-41470 Supplier Medchemexpress LLC Supplier No. HY107434S1MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Molindone-d8 is the deuterium-labeled isotopologue of molindone supplied as a research-grade stable isotope standard for analytical and preclinical studies. It is intended for use as an internal standard and tracer in mass spectrometry, metabolic, and pharmacokinetic research.

  • Deuterium-labeled analog for isotope-dilution assays.
  • High purity suitable for analytical use (99.2%).
  • Molecular formula C16H16D8N2O2 and molecular weight 284.42 g·mol⁻¹.
  • Offered in small quantities suitable for trace-level studies (1 mg, 5 mg, 10 mg).
  • Recommended storage: powder -20 °C for long term; in solvent -80 °C for short term.
  • Common applications: LC-MS/MS internal standard, metabolic tracing, and pharmacokinetic studies.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.