Learn More
Selleck Chemical LLC Molindone hydrochloride
SDP
Catalog No. 5000003247
Encompass_Preferred
Molindone hydrochloride is the hydrochloride salt form of molindone a conventional antipsychotic used in the therapy of schizophrenia It is a dopamine D2/D5 receptor antagonist
Catalog No. 50-000-03247 Supplier Selleck Chemical LLC Supplier No. S548125MG
Molindone hydrochloride is the hydrochloride salt form of molindone a conventional antipsychotic used in the therapy of schizophrenia It is a dopamine D2/D5 receptor antagonist