Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Molnupiravir | 00-00-0 | MFCD32663515 | 99.9% | 336.35 g/mol | C13H12D7N3O7 | 5 MG
SDP

Catalog No. 5000223505
Encompass_Preferred

Molnupiravir-d7 is the deuterium (d7) isotopologue of molnupiravir used as an analytical standard and tracer in antiviral research. It is intended for use as an internal standard in quantitative assays and to support pharmacokinetic, metabolic, and stability studies involving molnupiravir.

  • Deuterium-labeled (d7) isotopologue for tracer studies.
  • Molecular formula C13H12D7N3O7; molecular weight 336.35 g/mol.
  • High purity suitable for analytical applications (≈99.9%).
  • Suitable as an internal standard for quantitative LC-MS/MS assays.
  • Supports pharmacokinetic and metabolic profiling of antiviral compounds.
  • Available in small milligram quantities for research use.

Catalog No. 50-002-23505 Supplier Medchemexpress LLC Supplier No. HY135853S5MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Molnupiravir-d7 is the deuterium (d7) isotopologue of molnupiravir used as an analytical standard and tracer in antiviral research. It is intended for use as an internal standard in quantitative assays and to support pharmacokinetic, metabolic, and stability studies involving molnupiravir.

  • Deuterium-labeled (d7) isotopologue for tracer studies.
  • Molecular formula C13H12D7N3O7; molecular weight 336.35 g/mol.
  • High purity suitable for analytical applications (≈99.9%).
  • Suitable as an internal standard for quantitative LC-MS/MS assays.
  • Supports pharmacokinetic and metabolic profiling of antiviral compounds.
  • Available in small milligram quantities for research use.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.