Learn More
Apexbio Technology LLC Molnupiravir(Synonyms: EIDD-2801, MK-4482, Lagevrio, Molnupex, EIDD-1931-isopropyl ester, EIDD-2801 prodrug), 200mg, CAS: 2349386-89-4.
SDP
Catalog No. 5000061020
Encompass_Preferred
Molnupiravir (CAS 2349386-89-4) is an orally bioavailable nucleoside analog prodrug metabolized intracellularly into an active nucleoside triphosphate derivative Its antiviral effect occurs once incorporated by viral RNA-dependent RNA polymerase into newly synthesized viral RNA inducing an accumulation of mutations that disrupt viral genome fidelity and replication In research applications Molnupiravir is extensively employed for in vitro and in vivo studies investigating antiviral activities replication mechanisms and pharmacological responses especially against RNA viruses such as SARS-CoV-2 influenza virus and other coronaviruses
Catalog No. 50-000-61020 Supplier Apexbio Technology LLC Supplier No. B8578200
Molnupiravir (CAS 2349386-89-4) is an orally bioavailable nucleoside analog prodrug metabolized intracellularly into an active nucleoside triphosphate derivative Its antiviral effect occurs once incorporated by viral RNA-dependent RNA polymerase into newly synthesized viral RNA inducing an accumulation of mutations that disrupt viral genome fidelity and replication In research applications Molnupiravir is extensively employed for in vitro and in vivo studies investigating antiviral activities replication mechanisms and pharmacological responses especially against RNA viruses such as SARS-CoV-2 influenza virus and other coronaviruses