Learn More
Medchemexpress LLC Molnupiravir | 2492423-29-5 | 99.9% | 329.31 | 200 MG
SDP
Catalog No. 5000366051
Encompass_Preferred
Molnupiravir (EIDD-2801) is an orally bioavailable prodrug of the ribonucleoside analog EIDD-1931. It exhibits broad-spectrum antiviral activity against influenza virus and multiple coronaviruses, including SARS-CoV-2, MERS-CoV, and SARS-CoV. It is being researched for its potential use in COVID-19 and seasonal/pandemic influenza.
  • Orally bioavailable prodrug
  • Exhibits broad-spectrum antiviral activity
  • Effective against SARS-CoV-2, MERS-CoV, and SARS-CoV
  • Being researched for use in COVID-19 and influenza
  • Prevents SARS-CoV replication in C57BL/6 mice
  • Reduces shed virus load and fever duration in ferrets
Catalog No. 50-003-66051 Supplier Medchemexpress LLC Supplier No. 200MGHY135853
Molnupiravir (EIDD-2801) is an orally bioavailable prodrug of the ribonucleoside analog EIDD-1931. It exhibits broad-spectrum antiviral activity against influenza virus and multiple coronaviruses, including SARS-CoV-2, MERS-CoV, and SARS-CoV. It is being researched for its potential use in COVID-19 and seasonal/pandemic influenza.
  • Orally bioavailable prodrug
  • Exhibits broad-spectrum antiviral activity
  • Effective against SARS-CoV-2, MERS-CoV, and SARS-CoV
  • Being researched for use in COVID-19 and influenza
  • Prevents SARS-CoV replication in C57BL/6 mice
  • Reduces shed virus load and fever duration in ferrets