Learn More
ATCC Moloney/4070A MuLV; Strain: pAMS
Supplier:  ATCC VR1450
Encompass
Hybrid Moloney/Amphotropic murine leukemia virus (Mo/A-MuLv) 4070A envelope strain is propagated in NIH/3T3 cells (ATCC CRL-1658). This strain was harvested from tk(-) NIH3T3 cells transfected with molecular clone of MLV cells (ATCC VR-1448). Use this virus in your analytical methodologies and pharmaceutical and personal care applications.
Catalog No. 50-238-3690