Learn More
Cayman Chemical Molsidomine, Purity: ≥99%, CAS: 25717-80-0, 100 mg
SDP
Catalog No. 502892460
Encompass_Preferred
Molsidomine is a NO-releasing prodrug. Liver esterases convert molsidomine to the active metabolite, SIN-1 (half-life in plasma is 1-2 hours), which then releases NO.
Catalog No. 50-289-2460 Supplier Cayman Chemical Supplier No. 82200100 MG
Molsidomine is a NO-releasing prodrug. Liver esterases convert molsidomine to the active metabolite, SIN-1 (half-life in plasma is 1-2 hours), which then releases NO.