Learn More
ATCC MOLT-4; Leukemia; Human (Homo sapiens)
Catalog No. 502384190
Encompass
MOLT-4 is a T lymphoblast cell line derived from the same patient as the MOLT-3 cell line (ATCC CRL-1552). The line was established from cells taken from a 19-year-old, male patient with Acute lymphoblastic leukemia (ALL) in relapse. This cell line can be used in immunology and immune system disorder research.
Catalog No. 50-238-4190 Supplier ATCC Supplier No. CRL1582
MOLT-4 is a T lymphoblast cell line derived from the same patient as the MOLT-3 cell line (ATCC CRL-1552). The line was established from cells taken from a 19-year-old, male patient with Acute lymphoblastic leukemia (ALL) in relapse. This cell line can be used in immunology and immune system disorder research.