Learn More
Hach Company Molybdate 3 Reagent, 100mL
Supplier: Hach Company 199532
For low and ultra-low range silica determination by the Heteropoly Blue method. Hach methods 8282 and 8186. 100 mL Marked Dropping Bottle