Learn More
Hach Company Molybdate 3 Reagent, 500 mL
Supplier:  Hach Company 199549
Encompass
For low and ultra-low range silica determination by the Heteropoly Blue method. Hach methods 8282 and 8186. 500 mL bottle
Catalog No. 50-199-4652