Learn More
Hach Company Molybdate 3 Reagent Solution, 50 mL SCDB
Supplier:  Hach Company 199526
Encompass
For low range silica determination by the Heteropoly Blue method. 50 mL Self-Contained Dropping Bottle
Catalog No. 50-199-4650