Learn More
Hach Company Molybdate Reagent, MFCD00240888, 1 L
Supplier:  Hach Company 223653
Encompass
For high range phosphate (phosphorus) determination by the amino acid method (Hach method 8178). Range: Up to 30 mg/L PO4 1 liter bottle.
Catalog No. 50-465-378