Learn More
Hach Company Molybdate Reagent, 100 mL MDB
Supplier:  Hach Company 223632
Encompass
For high range phosphate (phosphorus) determination by the amino acid method (Hach method 8178). Range: Up to 30 mg/L PO₄ 100 mL Marked Dropping Bottle
Catalog No. 50-199-4741