Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CHEMetrics™ by AquaPhoenix Molybdate Refill Kit, Use with K-6720
SDP

Catalog No. 13299666
Change view
Click to view available options
Quantity:
30/Box
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
13-299-666 30/Box
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 13-299-666 Supplier CHEMetrics™ by AquaPhoenix Supplier No. R6720
Add to Cart
Add to Cart

Refill kit for molybdate water treatment test kits using self-filling ampoules with pre-measured reagent. Compare results to supplied color standards to determine concentration.

  • This product is a refill for K-6720 Molybdate CHEMets Visual Kit for the analysis of molybdate in water.
  • Contains 30 self-filling CHEMets ampoules packaged securely in cardboard trays and box.

Specifications

pH Range 7
Volume (Metric) 0.5 mL
For Use With (Equipment) K-6720
No. of Tests 30
Quantity 30/Box
Type Molybdate Refill Kit
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.