Learn More
Hach Company Molybdate Reagent Powder Pillows for HR Silica, 25mL, pk100
Catalog No. 501994496 Shop All Hach Company Products
Encompass
For determination of high range silica by the Silicomolybdate method. Hach method 8185. Sample size: 25 mL. Pack of 100 powder pillows
Catalog No. 50-199-4496 Supplier Hach Company Supplier No. 104199
For determination of high range silica by the Silicomolybdate method. Hach method 8185. Sample size: 25 mL. Pack of 100 powder pillows