Learn More
Hach Company Molybdate Reagent Powder Pillows for LR Silica, 5 mL, pk/100
Catalog No. 501995067 Shop All Hach Company Products
Encompass
For determination of low range silica (0-2 mg/L). 5-mL sample. For use with Hach visual test kits.
Catalog No. 50-199-5067 Supplier Hach Company Supplier No. 1454699
For determination of low range silica (0-2 mg/L). 5-mL sample. For use with Hach visual test kits.