Learn More
Hach Company Molybdate Reagent Solution for Phosphorus, 500 mL
Supplier:  Hach Company 2599849
Encompass
For determination of low range Reactive Phosphorus (Phosphate) by the Rapid Liquid Ascorbic Acid Method. Hach Method 10055
Catalog No. 50-199-6303