Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CHEMetrics™ by AquaPhoenix Molybdate Water Quality Visual Test Kit, 2 to 24 ppm as Mo
SDP

Catalog No. 13299653
Change view
Click to view available options
No. of Tests:
30
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. No. of Tests
13-299-653 30
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 13-299-653 Supplier CHEMetrics™ by AquaPhoenix Supplier No. K6702
Add to Cart
Add to Cart

Molybdate visual test kit featuring self-filling ampoules with pre-measured reagent for a single test. Compare results to supplied color standards to determine concentration.

  • Kit comes in a plastic case and contains everything needed to perform 30 tests: Refill, Comparator, 25 mL sample cup, and instructions.

Specifications

Analytes Molybdate
pH Range 7
Volume (Metric) 0.5 mL
No. of Tests 30
Range 2 to 24 ppm
Type Water Quality Visual Test Kit
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.