Learn More
Electron Microscopy Sciences Omniprobe FIB Lift-Out Grid 4-Post
SDP
Catalog No. 5033767
Encompass
Custom 4-post lift-out grids for in-situ lift-out. Include multiple indexed mounting locations, two with vertical bars attachment surfaces and two with "V" shaped alignment surfaces. Sides have lower profile for easier access to outermost posts. 3 mm diameter.
Catalog No. 50-337-67 Supplier Electron Microscopy Sciences Supplier No. 7596406
Custom 4-post lift-out grids for in-situ lift-out. Include multiple indexed mounting locations, two with vertical bars attachment surfaces and two with "V" shaped alignment surfaces. Sides have lower profile for easier access to outermost posts. 3 mm diameter.