Learn More
Hach Company Molybdenum 1 Reagent Powder Pillows, 50 mL, pk/100
Supplier:  Hach Company 2352769
Encompass
For determination of low range molybdenum by the ternary complex method (Hach method 8169). Range: 0.02-3.00 mg/L Mo Sample Size: 50 mL
Catalog No. 50-199-5866