Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Description
Molybdenum
Spécifications
Spécifications
| Melting Point | 2617°C |
| CAS | 7439-98-7 |
| Boiling Point | 4612°C |
| Density | 10.3 g/mL (at 25°C) |
| Quantity | 1 Ea. |
| Molecular Formula | Mo |
| Linear Formula | Mo |
| MDL Number | MFCD00003465 |
| Synonym | Molybdenum; MO007920 |
| Molecular Weight (g/mol) | 95.94 |
| Afficher plus de résultats |
missing translation for 'productTitle'
missing translation for 'byClickingSubmit' missing translation for 'privacyPolicy'.
missing translation for 'spotOppurtunityImprovement'