Learn More
Hach Company Molybdenum 2 Reagent Solution, 100 mL MDB
Supplier:  Hach Company 2352532
Encompass
For determination of low range molybdenum by the ternary complex method (Hach method 8169). Range: 0.02-3.00 mg/L Mo
Catalog No. 50-199-5864