Learn More
Hach Company Molybdenum 2 Reagent Solution, 25 mL SCDB
Supplier:  Hach Company 2352523
Encompass
For determination of low range molybdenum by the ternary complex method (Hach method 8169). Range: 0.02-3.00 mg/L Mo
Catalog No. 50-199-5863