Learn More
Strem, An Ascensus Company CAS# 12069-89-5. 50g. Molybdenum carbide (99.5%-Mo). MFCD00014218
SDP
Catalog No. 5090115338
Encompass
CAS# 12069-89-5. 50g. Molybdenum carbide (99.5%-Mo). MFCD00014218. Molecular Weight: 203.91. Molecular Formula: Mo2C. Color/form: gray pwdr. Strem# 93-4243. http://www.strem.com/catalog/v/93-4243/
Catalog No. 50-901-15338 Supplier Strem, An Ascensus Company Supplier No. 93424350G
CAS# 12069-89-5. 50g. Molybdenum carbide (99.5%-Mo). MFCD00014218. Molecular Weight: 203.91. Molecular Formula: Mo2C. Color/form: gray pwdr. Strem# 93-4243. http://www.strem.com/catalog/v/93-4243/