Learn More
Strem, An Ascensus Company CAS # 13939-06-5. Molybdenum carbonyl, 98%. MFCD00003466. 100G
SDP
Supplier:  Strem, An Ascensus Company 421350100G
Encompass
CAS# 13939-06-5. 100G. Molybdenum carbonyl, 98%. MFCD00003466. Molecular Formula: C6MoO6 Molecular Weight: 264.01. Color/form white xtl. Strem#: 42-1350. www.strem.com/catalog/v3/42-1350/
Catalog No. 50-901-11704