Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Molybdenum disulfide nanoplatelets

Catalog No. 901792250MG Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
250 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
90-179-2250MG 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 90-179-2250MG Supplier Sigma Aldrich Supplier No. 901792250MG
Only null left
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Molybdenum disulfide nanoplatelet (MoS2) is a two dimensional layered transition metal dichalcogenide (TMD) which has hexagonal layers of molybdenum atoms that are sandwiched between two layers of chalcogen atoms. It can be used as a non-linear optical material and as a reinforcing filler in polymeric composites.Owing to the thickness dependent physical and chemical properties, the transition metal dichalcogenide (TMD) based two dimensional (2D) nanomaterials have drawn significant attention recently. The atomically thin layers of MoS2 and WS2 represent important class of TMD based 2D-material and they find applications in photocatalysis, phtotovoltaics, gas sensing, lithium ion batteries, field effect transistors, spintronics etc.

Specifications

Linear Formula MoS2
Quantity 250 mg
Synonym 2 D-Moly sulfide; 2 D-Molybdenum disulfide; 2 D-Molybdenum disulfide with nonionic surfactant; Moly sulfide
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.