Learn More
Sigma Aldrich Fine Chemicals Biosciences Molybdenum foil, 15mm disks, thickness 0.1mm, annealed, 99.9% | 7439-98-7 | MFCD00003465 | 10EA
Catalog No. 501850257
Encompass
Molybdenum foil, 15mm disks, thickness 0.1mm, annealed, 99.9% | Purity: 99.9% | Mol Wt: 95.94 | 7439-98-7 | MFCD00003465 | 10EA
Catalog No. 501850257 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. GF7792735010EA
Molybdenum foil, 15mm disks, thickness 0.1mm, annealed, 99.9% | Purity: 99.9% | Mol Wt: 95.94 | 7439-98-7 | MFCD00003465 | 10EA