Learn More
Sigma Aldrich Fine Chemicals Biosciences Molybdenum foil, light tested, 25x25mm, thickness 0.01mm, annealed, 99.9% | 7439-98-7 | MFCD00003465 | 1EA
Catalog No. 501850303
Encompass
Molybdenum foil, light tested, 25x25mm, thickness 0.01mm, annealed, 99.9% | Purity: 99.9% | Mol Wt: 95.94 | 7439-98-7 | MFCD00003465 | 1EA
Catalog No. 501850303 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. GF808157761EA
Molybdenum foil, light tested, 25x25mm, thickness 0.01mm, annealed, 99.9% | Purity: 99.9% | Mol Wt: 95.94 | 7439-98-7 | MFCD00003465 | 1EA