Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Molybdenum gauze, 35 mesh woven from 0.0509mm (0.002 in.) dia. wire, Thermo Scientific™

Catalog No. AA42485FL
Change view
Click to view available options
75x75mm
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
AA42485FL 75x75mm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AA42485FL Supplier Thermo Scientific Chemicals Supplier No. 42485FL

missing translation for 'validMainframeMsg'

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Specifications

CAS 7439-98-7
Molecular Formula Mo
MDL Number MFCD00003465
Quantity 75 x 75mm
Synonym molybdenum, elemental, 6+ ion, mchvl, vi ion, molybdenum, metallic, metco 63, atom, molybdenum, ion 6+, tsm1, amperit 106.2
InChI Key ZOKXTWBITQBERF-UHFFFAOYSA-N
SMILES [Mo]
IUPAC Name molybdenum
Molecular Weight (g/mol) 95.95
PubChem CID 23932
ChEBI CHEBI:28685
Mesh Size 35 Mesh
Form Wire Cloth
Chemical Name or Material Molybdenum gauze, 35 mesh woven from 0.0509mm (0.002 in.) dia. wire
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.