Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Molybdenum hexacarbonyl, 98%, ACROS Organics™

Click to view available options
Quantity:
10g
50g
250g
Packaging:
Glass bottle
13939-06-5
C6O6Mo
264
MFCD00003466
KMKBZNSIJQWHJA-UHFFFAOYSA-N
molybdenumhexacarbonyl, carbon monooxide-molybdenum 6:1, hexakis carbon monoxide molybdenum, hexacarbonylmolybdenu, hexakis methanidylidyneoxidanium molybdenum, hexacarbonylmolybdenum, may contain up to cr 5g
2724068
CHEBI:30508
carbon monoxide;molybdenum
[C-]#[O+].[C-]#[O+].[C-]#[O+].[C-]#[O+].[C-]#[O+].[C-]#[O+].[Mo]
13939-06-5
C6O6Mo
264
MFCD00003466
KMKBZNSIJQWHJA-UHFFFAOYSA-N
molybdenumhexacarbonyl, carbon monooxide-molybdenum 6:1, hexakis carbon monoxide molybdenum, hexacarbonylmolybdenu, hexakis methanidylidyneoxidanium molybdenum, hexacarbonylmolybdenum, may contain up to cr 5g
2724068
CHEBI:30508
carbon monoxide;molybdenum
[C-]#[O+].[C-]#[O+].[C-]#[O+].[C-]#[O+].[C-]#[O+].[C-]#[O+].[Mo]

Molybdenum hexacarbonyl, 98%, C6O6Mo, CAS Number-13939-06-5, hexacarbonylmolybdenu, carbon monooxide-molybdenum 6:1, hexacarbonylmolybdenum, may contain up to cr 5g, hexakis carbon monoxide molybdenum, molybdenumhexacarbonyl, hexakis methanidylidyneoxidanium molybdenum, 50g, 100.0, 97.5, CHEBI:30508

Molybdenum hexacarbonyl, 98%, C6O6Mo, CAS Number-13939-06-5, hexacarbonylmolybdenu, carbon monooxide-molybdenum 6:1, hexacarbonylmolybdenum, may contain up to cr 5g, hexakis carbon monoxide molybdenum, molybdenumhexacarbonyl, hexakis methanidylidyneoxidanium molybdenum, 50g, 100.0, 97.5, CHEBI:30508

Color White to Yellow, White through Yellow
CAS Min % 97.5
CAS Max % 100.0
Assay Percent Range 98%
Linear Formula Mo(CO)6
Fieser 02,287; 03,206; 04,346; 09,317; 10,274; 13,194; 01,140; 02,448; 03,325; 04,559; 05,736; 06,652; 07,68; 08,109; 09,113; 15,9
Solubility Information Solubility in water: insoluble. Other solubilities: soluble in benzene, slightly soluble in ether and most oter common, organic solvents
Formula Weight 264
Physical Form Crystalline Powder
Percent Purity 98%
Density 1.9600g/mL
Elemental Analysis 97.5% min.
Infrared Spectrum Authentic
Packaging Glass bottle
Solubility Solubility in water: insoluble. Other solubilities: soluble in benzene, slightly soluble in ether and most oter common, organic solvents
Specific Gravity 1.96
Melting Point 150.0°C
Quantity 250g
Chemical Name or Material Molybdenum hexacarbonyl, 98%
Show More Show Less

GHS H Statement
Fatal if swallowed.
Fatal in contact with skin.
Fatal if inhaled.

GHS P Statement
Rinse mouth.
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Gently wash with plenty of soap and water.
IF INHALED: Remove to fresh air and keep at rest in a position comfortabl

GHS Signal Word: Danger

EINECSNumber : 237-713-3

TSCA : TSCA