Learn More
Crescent Chemical Co Inc MOLYBDENUM ICP STD 500ML
SDP
Catalog No. 5072036
Encompass_Preferred
Molybdenum, NH40H Single Element Plasma Emission Spectroscopy Standards 1,000 UgmL NIST Traceable CAS Size - 500mL Catalog Number - 8568.5
Catalog No. 50-720-36 Supplier Crescent Chemical Co Inc Supplier No. IS8568 (500ML)
Molybdenum, NH40H Single Element Plasma Emission Spectroscopy Standards 1,000 UgmL NIST Traceable CAS Size - 500mL Catalog Number - 8568.5