Learn More
Strem, An Ascensus Company CAS# 13478-18-7. 1g. Molybdenum(III) chloride (99.5%-Mo). MFCD00014214
SDP
Catalog No. 5090115327
Encompass
CAS# 13478-18-7. 1g. Molybdenum(III) chloride (99.5%-Mo). MFCD00014214. Molecular Weight: 202.32. Molecular Formula: MoCl3. Color/form: -100 mesh dark red-brown solid Strem# 93-4214. http://www.strem.com/catalog/v/93-4214/
Catalog No. 50-901-15327 Supplier Strem, An Ascensus Company Supplier No. 9342141G
CAS# 13478-18-7. 1g. Molybdenum(III) chloride (99.5%-Mo). MFCD00014214. Molecular Weight: 202.32. Molecular Formula: MoCl3. Color/form: -100 mesh dark red-brown solid Strem# 93-4214. http://www.strem.com/catalog/v/93-4214/