Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Molybdenum(IV) sulfide

Catalog No. 80416910G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Change view
Click to view available options
Quantity:
10 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
80-416-910G 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 80-416-910G Supplier Sigma Aldrich Supplier No. 80416910G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

MoS2 is an important compound with applications in batteries, lubricants and hydrogen production. The layered structure of MoS2 allows single layers of the compound to be used as 2D nanosheets similar to graphene.

Specifications

CAS 1317-33-5
Quantity 10 g
Molecular Formula MoS2
Linear Formula MoS2
MDL Number MFCD00003470
Synonym Molybdenum disulfide
Molecular Weight (g/mol) 160.07
Percent Purity 99% trace metals basis
Density 5.06 g/mL (at 25°C)
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.