Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Molybdenum(IV) sulfide

Catalog No. 234842100G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
23-484-2100G 100 g
23-484-2500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 23-484-2100G Supplier Sigma Aldrich Supplier No. 234842100G
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

We are committed to bringing you Greener Alternative Products, which adhere to one or more of The 12 Principles of Greener Chemistry. This product has been enhanced for energy efficiency. Find details here.Hydrogenation and isomerization catalyst. Photoresponse of MoS2 nanosheets, formed by chemical route was studied. MoS2 may find potential applications in electronics and optoelectronics.

Specifications

CAS 1317-33-5
Quantity 100 g
Molecular Formula MoS2
Linear Formula MoS2
MDL Number MFCD00003470
Synonym Molybdenum disulfide
Molecular Weight (g/mol) 160.07
Percent Purity 98%
Density 5.06 g/mL (at 25°C)
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.