Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Molybdenum(IV) sulfide

Catalog No. 69860500G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Change view
Click to view available options
Quantity:
100 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
69-860-500G 500 g
69-860-100G 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 69-860-500G Supplier Sigma Aldrich Supplier No. 69860500G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Molybdenum disulfide is a two dimensional layered material. Monolayers of transition metal dichalcogenides (TMDs)exhibit photoconductivity. The layers of the TMD can be mechanically or chemicaly exfoliated to form nanosheets.Acidic solution of MoS2 particles was used to catalyze hydrogen evolution at a water 1,2-dichloroethane interface. MoS2 was dispersed in N-methyl-pyrrolidone to form exfoliated MoS2 flakes of various sizes. MoS2 ink was used for inkjet printer. MoS2 may find potential applications in electronics and optoelectronics.

Specifications

CAS 1317-33-5
Quantity 500 g
Molecular Formula MoS2
Linear Formula MoS2
MDL Number MFCD00003470
Synonym Molybdenum disulfide
Molecular Weight (g/mol) 160.07
Density 5.06 g/mL (at 25°C)
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.