Learn More
Molybdenum(IV) sulfide, 98%, Thermo Scientific Chemicals
Description
It is widely used as a solid lubricant .It finds applications mainly as Intermediates, Process regulators, Processing aids. Employed as a co catalyst for desulfurization in petrochemistry. As a catalyst in the electrolysis of water.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Melting Point | 1185°C |
Color | Black |
Physical Form | Powder |
Quantity | 1 kg |
Assay Percent Range | 98% |
Solubility Information | Insoluble in water and dilute acids |
IUPAC Name | bis(sulfanylidene)molybdenum |
Formula Weight | 160.08 |
Odor | Odorless |
Appearance | Black powder |
Show More |
Safety and Handling
Hazard Category | H332 |
Precautionary Statement | P261-P271-P304+P340-P312 |
EINECSNumber | 215-263-9 |
RTECSNumber | QA4697000 |
TSCA | Yes |
Recommended Storage | Ambient temperatures |
RUO – Research Use Only
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.