Learn More
Sigma Aldrich Fine Chemicals Biosciences Molybdenum mesh, 50x50mm, nominal aperture 0.8mm, wire diameter 0.18mm, 26 wires/inch, open area 67%, plain weave mesh | 7439-98-7 | MFCD00003465 | 5EA
Catalog No. 501849769
Encompass
Molybdenum mesh, 50x50mm, nominal aperture 0.8mm, wire diameter 0.18mm, 26 wires/inch, open area 67%, plain weave mesh | Mol Wt: 95.94 | 7439-98-7 | MFCD00003465 | 5EA
Catalog No. 501849769 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. GF400158315EA
Molybdenum mesh, 50x50mm, nominal aperture 0.8mm, wire diameter 0.18mm, 26 wires/inch, open area 67%, plain weave mesh | Mol Wt: 95.94 | 7439-98-7 | MFCD00003465 | 5EA