Learn More
Sigma Aldrich Fine Chemicals Biosciences Molybdenum microfoil, disks, 25mm, thinness 0.1mum, specific density 92.4mug/cm2, permanent mylar 3.5mum support, 99.9% | 7439-98-7 | MFCD00003465 | 5EA
Catalog No. 501850224
Encompass
Molybdenum microfoil, disks, 25mm, thinness 0.1mum, specific density 92.4mug/cm2, permanent mylar 3.5mum support, 99.9% | Purity: 99.9% | Mol Wt: 95.94 | 7439-98-7 | MFCD00003465 | 5EA
Catalog No. 501850224 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. GF755745925EA
Molybdenum microfoil, disks, 25mm, thinness 0.1mum, specific density 92.4mug/cm2, permanent mylar 3.5mum support, 99.9% | Purity: 99.9% | Mol Wt: 95.94 | 7439-98-7 | MFCD00003465 | 5EA