Learn More
Strem, An Ascensus Company CAS# 7439-98-7. 100g. Molybdenum powder (99.95%). MFCD00003465
SDP
Catalog No. 5090115352
Encompass
CAS# 7439-98-7. 100g. Molybdenum powder (99.95%). MFCD00003465. Molecular Weight: 95.94. Molecular Formula: Mo. Color/form: -100 mesh Strem# 93-4256. http://www.strem.com/catalog/v/93-4256/
Catalog No. 50-901-15352 Supplier Strem, An Ascensus Company Supplier No. 934256100G
CAS# 7439-98-7. 100g. Molybdenum powder (99.95%). MFCD00003465. Molecular Weight: 95.94. Molecular Formula: Mo. Color/form: -100 mesh Strem# 93-4256. http://www.strem.com/catalog/v/93-4256/