Learn More
Molybdenum Standard, For AAS, 1,000 mg/L Mo in Hydrochloric Acid, MilliporeSigma™ Supelco™
Shop All MilliporeSigma Supelco ProductsSpecifications
Specifications
| Product Line | TraceCERT |
| Concentration | 1000 mg/L Mo in hydrochloric acid |
| Quantity | 250 mL |
| Grade | TraceCERT™ Analytical Standard |
TraceCERT is a registered trademark of Sigma-Aldrich Co. LLC
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.