Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MP Biomedicals™ Molybdenum Trioxide, ≥99.5%, (ACS Reagent Grade), MP Biomedicals

Catalog No. ICN15254890 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
100 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN15254890 500 g
ICN15254880 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. ICN15254890 Supplier MP Biomedicals™ Supplier No. 0215254890
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

CAS: 1313-27-5 | MoO3 | 143.95 g/mol

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1313-27-5
Molecular Formula MoO3
Molecular Weight (g/mol) 143.95
MDL Number MFCD00003469
InChI Key JKQOBWVOAYFWKG-UHFFFAOYSA-N
Synonym molybdenum trioxide, molybdenum vi oxide, molybdic anhydride, molybdic trioxide, molybdena, molybdic oxide, molybdenum oxide, molybdic acid anhydride, molybdenum peroxide
PubChem CID 14802
ChEBI CHEBI:30627
IUPAC Name trioxomolybdenum
SMILES O=[Mo](=O)=O

Specifications

Color Yellow
Quantity 500 g
Assay Percent Range ≥99.5%
Linear Formula MoO3
Formula Weight 143.94
Grade ACS Reagent
Concentration or Composition (by Analyte or Components) Assay (MoO3) - >99.5%, Insoluble Matter in dilute NH4OH - <0.0100%, Ammonium (NH4) - <0.0020%, Arsenate and Silicate (as SiO2) - <0.0010%, Chloride (Cl) - <0.0020%, Heavy Metals (as Pb)- <0.0050%, Nitrate (NO3)- <0.0030%, Phosphate (PO4)- <0.0005%, Sulfate (SO4) - <0.0200%
Chemical Name or Material Molybdenum Trioxide
Recommended Storage Room Temperature
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.