Learn More
Sigma Aldrich Fine Chemicals Biosciences Molybdenum tube, 100mm, outside diameter 0.5mm, inside diameter 0.38mm, wall thickness 0.06mm, 99.9% | 7439-98-7 | MFCD00003465 | 3EA
Catalog No. 501850247
Encompass
Molybdenum tube, 100mm, outside diameter 0.5mm, inside diameter 0.38mm, wall thickness 0.06mm, 99.9% | Purity: >=99.9% | Mol Wt: 95.94 | 7439-98-7 | MFCD00003465 | 3EA
Catalog No. 501850247 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. GF765725133EA
Molybdenum tube, 100mm, outside diameter 0.5mm, inside diameter 0.38mm, wall thickness 0.06mm, 99.9% | Purity: >=99.9% | Mol Wt: 95.94 | 7439-98-7 | MFCD00003465 | 3EA