Learn More
Sigma Aldrich Fine Chemicals Biosciences Molybdenum tube, 200mm, outside diameter 6.0mm, inside diameter 5.6mm, wall thickness 0.20mm, 99.9% | 7439-98-7 | MFCD00003465 | 5EA
Catalog No. 501850267
Encompass
Molybdenum tube, 200mm, outside diameter 6.0mm, inside diameter 5.6mm, wall thickness 0.20mm, 99.9% | Purity: 99.9% | Mol Wt: 95.94 | 7439-98-7 | MFCD00003465 | 5EA
Catalog No. 501850267 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. GF781558715EA
Molybdenum tube, 200mm, outside diameter 6.0mm, inside diameter 5.6mm, wall thickness 0.20mm, 99.9% | Purity: 99.9% | Mol Wt: 95.94 | 7439-98-7 | MFCD00003465 | 5EA